3'-Methoxyacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 3′-Methoxyacetophenone

A139173-100G A139173-10G A139173-25G A139173-500G
77272-398EA 28.84 USD
77272-398 77272-400 77272-402 77272-404
3'-Methoxyacetophenone 97%
3'-Methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00008736
CAS Number: 586-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR