N-(3-Bromo-5-chloropyridin-2-yl)formamide 97%

Supplier: Ambeed
Warning

A139167-100MG A139167-250MG
77234-844EA 5.15 USD
77234-844 77234-846
N-(3-Bromo-5-chloropyridin-2-yl)formamide 97%
N-(3-Bromo-5-chloropyridin-2-yl)formamide

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O
MW: 235.47 g/mol
MDL Number: MFCD18324138
CAS Number: 1263214-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR