N-(4-Methoxy-2-methyl-6-nitrophenyl)acetamide 97%

Supplier: Ambeed
Warning

A139148-1G A139148-250MG
77234-840EA 26.78 USD
77234-840 77234-842
N-(4-Methoxy-2-methyl-6-nitrophenyl)acetamide 97%
N-(4-Methoxy-2-methyl-6-nitrophenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₄
MW: 224.21 g/mol
MDL Number: MFCD28975071
CAS Number: 500562-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR