5-Amino-α,α,α-trifluoro-m-toluic acid 97%
Supplier: Ambeed
|
Warning
|
A139102-100G
A139102-10G
A139102-1G
A139102-250MG
A139102-25G
A139102-5G
77234-828EA
386
USD
77234-828
77234-830
77234-832
77234-834
77234-836
77234-838
5-Amino-α,α,α-trifluoro-m-toluic acid 97%
5-Amino-α,α,α-trifluoro-m-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₆F₃NO₂ |
MDL Number:
MFCD00236641 CAS Number: 328-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...