3-Amino-2-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Nitro-3-pyridylamine, 2-Nitropyridin-3-amine, 2-Nitropyridin-3-ylamine

A139101-100G A139101-10G A139101-1G A139101-25G A139101-5G
77285-648EA 122 USD
77285-648 77285-650 77285-652 77285-654 77285-656
3-Amino-2-nitropyridine 98%
3-Amino-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD00044102
CAS Number: 13269-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR