α,α,α,2,4-Pentafluorotoluene 98%

Supplier: Ambeed
Warning

A138916-1G A138916-25G A138916-5G
77246-820EA 7 USD
77246-820 77246-822 77246-824
α,α,α,2,4-Pentafluorotoluene 98%
α,α,α,2,4-Pentafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃F₅
MDL Number: MFCD04972808
CAS Number: 64248-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR