1-bromo-2,4-dimethyl-5-nitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,4-dimethyl-5-nitrobenzene

A138877-10G A138877-250MG A138877-5G
77234-822EA 41.2 USD
77234-822 77234-824 77234-826
1-bromo-2,4-dimethyl-5-nitrobenzene 97%
1-bromo-2,4-dimethyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.061 g/mol
MDL Number: MFCD00075824
CAS Number: 69383-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR