Methyl 4-bromo-2-naphthoate 97%

Supplier: Ambeed
Warning

A138800-1G A138800-250MG
77258-740EA 849.75 USD
77258-740 77747-984
Methyl 4-bromo-2-naphthoate 97%
Methyl 4-bromo-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₂
MW: 265.10 g/mol
MDL Number: MFCD18413035
CAS Number: 5043-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR