4-Ethyl-4'-iodobiphenyl 95%

Supplier: Ambeed
Warning

A138785-100MG A138785-1G A138785-250MG
77285-632EA 40 USD
77285-632 77285-634 77285-636
4-Ethyl-4'-iodobiphenyl 95%
4-Ethyl-4'-iodobiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃I
MDL Number: MFCD03427551
CAS Number: 17078-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR