2,4-Dibromo-6-nitrophenol 95%

Supplier: Ambeed
Danger

A138727-1G A138727-250MG A138727-25G A138727-5G
77246-812EA 21.99 USD
77246-812 77246-814 77246-816 77246-818
2,4-Dibromo-6-nitrophenol 95%
2,4-Dibromo-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₃
MDL Number: MFCD00024232
CAS Number: 15969-09-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR