8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 95+%

Supplier: Ambeed
Warning

A138720-1G A138720-250MG A138720-5G
77246-806EA 75.19 USD
77246-806 77246-808 77246-810
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 95+%
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂
MW: 186.25 g/mol
MDL Number: MFCD05663532
CAS Number: 64172-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR