Methyl 4-amino-3,6-dichloropicolinate 97%

Supplier: Ambeed
Warning

A138609-1G A138609-5G
77234-818EA 110.21 USD
77234-818 77234-820
Methyl 4-amino-3,6-dichloropicolinate 97%
Methyl 4-amino-3,6-dichloropicolinate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
MDL Number: MFCD28722246
CAS Number: 350601-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR