2-Fluoro-p-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-anisidine, 2-Fluoro-4-methoxyaniline, 4-Amino-3-fluoroanisole

A138484-100G A138484-10G A138484-1G A138484-25G A138484-5G
77258-730EA 278 USD
77258-730 77258-732 77258-734 77258-736 77258-738
2-Fluoro-p-anisidine 98%
2-Fluoro-p-anisidine

Precaución: for Research Use Only

Formula: C₇H₈FNO
MDL Number: MFCD00047827
CAS Number: 458-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR