2,2'-O-Cyclouridine 98%

Supplier: Ambeed
Warning

A138388-100MG A138388-25G A138388-5G A138388-100G A138388-500G A138388-1000G
77272-358EA 5 USD
77272-358 77272-360 77272-362 77566-782 77615-100 77747-980
2,2'-O-Cyclouridine 98%
2,2'-O-Cyclouridine

Precaución: For research use only.

Formula: C₉H₁₀N₂O₅
MDL Number: MFCD00004945
CAS Number: 3736-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR