N-Boc-PEG3-bromide 97%

Supplier: Ambeed
Warning

Synonyms: Bocamine-PEG3-bromide, tert-Butyl (2-(2-(2-(2-bromoethoxy)ethoxy)ethoxy)ethyl)carbamate

A138365-100MG A138365-250MG A138365-1G A138365-5G A138365-25G
77234-814EA 17 USD
77234-814 77234-816 77315-628 77315-630 77745-982
N-Boc-PEG3-bromide 97%
N-Boc-PEG3-bromide

Precaución: for Research Use Only

Formula: C₁₃H₂₆BrNO₅
MDL Number: MFCD09840081
CAS Number: 1076199-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR