o-Anisaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde

A138178-100G A138178-10G A138178-25G A138178-500G A138178-1000G
77258-722EA 8.24 USD
77258-722 77258-724 77258-726 77258-728 77615-098
o-Anisaldehyde 98%
o-Anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00003308
CAS Number: 135-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR