3,5-Dichloro-4-methoxybenzenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-4-methoxybenzoic acid

A138095-1G A138095-250MG A138095-5G
77234-798EA 20 USD
77234-798 77234-800 77234-802
3,5-Dichloro-4-methoxybenzenecarboxylic acid 97%
3,5-Dichloro-4-methoxybenzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
MDL Number: MFCD00210404
CAS Number: 37908-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR