N-(4-Chloropyridin-3-yl)-4-((2,2-difluorobenzo[d][1,3]dioxol-5-yl)methyl)piperazine-1-carboxamide 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
JNJ-42165279
A138083-10MG
A138083-25MG
A138083-5MG
77234-792EA
49.44
USD
77234-792
77234-794
77234-796
N-(4-Chloropyridin-3-yl)-4-((2,2-difluorobenzo[d][1,3]dioxol-5-yl)methyl)piperazine-1-carboxamide 98+%
N-(4-Chloropyridin-3-yl)-4-((2,2-difluorobenzo[d][1,3]dioxol-5-yl)methyl)piperazine-1-carboxamide
Precaución: for Research Use Only
Formula:
C₁₈H₁₇ClF₂N₄O₃ MW: 410.8 g/mol |
MDL Number:
MFCD29047142 CAS Number: 1346528-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...