6,7-Dimethoxyquinoxaline-2,3(1H,4H)-dione 97%

Supplier: Ambeed
Warning

A137997-100MG A137997-250MG
77246-786EA 30.9 USD
77246-786 77246-788
6,7-Dimethoxyquinoxaline-2,3(1H,4H)-dione 97%
6,7-Dimethoxyquinoxaline-2,3(1H,4H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₄
MW: 222.2 g/mol
MDL Number: MFCD04038944
CAS Number: 4784-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR