4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 95%

Supplier: Ambeed
Warning

Synonyms: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol

A137942-100G A137942-25G A137942-500G A137942-5G
77246-776EA 10 USD
77246-776 77246-778 77246-780 77246-782 77246-784
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 95%
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL)

Precaución: for Research Use Only

Formula: C₉H₁₈NO₂
MW: 172.25 g/mol
MDL Number: MFCD00006478
CAS Number: 2226-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR