2,6-Difluoro-3-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A137516-1G A137516-25G A137516-5G
77246-762EA 13.39 USD
77246-762 77246-764 77246-766
2,6-Difluoro-3-methoxyphenylboronic acid 98%
2,6-Difluoro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
MDL Number: MFCD06657880
CAS Number: 870779-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR