4-Fluoro-N-methoxy-N-methylbenzamide 97%

Supplier: Ambeed
Warning

A137514-10G A137514-1G A137514-25G A137514-500G A137514-5G A137514-100G
77285-598EA 21.63 USD
77285-598 77285-600 77285-602 77285-604 77285-606 77747-973
4-Fluoro-N-methoxy-N-methylbenzamide 97%
4-Fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Number: MFCD02179265
CAS Number: 116332-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR