2-(Chloromethyl)benzoxazole 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Chloromethyl)benzo[d]oxazole, 2-(Chloromethyl)-1,3-benzoxazole

A137513-1G A137513-250MG A137513-5G
77234-758EA 56.65 USD
77234-758 77234-760 77234-762
2-(Chloromethyl)benzoxazole 95%
2-(Chloromethyl)benzoxazole

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.5900 g/mol
MDL Number: MFCD05664964
CAS Number: 41014-43-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR