(2R,4R)-4-Hydroxypiperidine-2-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A137473-100MG A137473-1G A137473-250MG
77246-756EA 132.87 USD
77246-756 77246-758 77246-760
(2R,4R)-4-Hydroxypiperidine-2-carboxylic acid hydrochloride 97%
(2R,4R)-4-Hydroxypiperidine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₃
MW: 181.61 g/mol
MDL Number: MFCD30541795
CAS Number: 133696-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR