Methanesulfonato[2-(dicyclohexylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

A137413-100MG A137413-1G A137413-250MG
77246-750EA 10 USD
77246-750 77246-752 77246-754
Methanesulfonato[2-(dicyclohexylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato[2-(dicyclohexylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₃₉H₄₉N₂O₃PPdS
MDL Number: MFCD28411649
CAS Number: 1445085-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR