2,5-Dibromothiophene 98%

Supplier: Ambeed
Warning

A137367-1000G A137367-100G A137367-10G A137367-500G A137367-25G
77285-590EA 47 USD
77285-590 77285-592 77285-594 77285-596 77315-404
2,5-Dibromothiophene 98%
2,5-Dibromothiophene

Precaución: for Research Use Only

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 211
Melting Pt: -6
MDL Number: MFCD00005420
CAS Number: 3141-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR