2-(Diphenylphosphino)ethylamine 95%

Supplier: Ambeed
Danger

A137345-250MG A137345-1G
77246-744EA 47 USD
77246-744 77745-975
2-(Diphenylphosphino)ethylamine 95%
2-(Diphenylphosphino)ethylamine

Precaución: for Research Use Only

Formula: (C₆H₅)₂PCH₂CH₂NH₂
MDL Number: MFCD00233831
CAS Number: 4848-43-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR