Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A137264-100MG A137264-1G A137264-250MG A137264-5G
77258-708EA 14.42 USD
77258-708 77258-710 77258-712 77549-670
Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate 97%
Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₄S
MW: 258.29 g/mol
MDL Number: MFCD16171042
CAS Number: 38067-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR