2-Chloro-4-fluoro-5-nitrophenol 97%

Supplier: Ambeed
Danger

A137158-1G A137158-5G
77285-576EA 7 USD
77285-576 77285-578
2-Chloro-4-fluoro-5-nitrophenol 97%
2-Chloro-4-fluoro-5-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₃
MDL Number: MFCD02670258
CAS Number: 84478-75-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR