Boc-Glu-OH 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Glu-OH, N-Boc-L-glutamic Acid

A137145-10G A137145-25G A137145-500G A137145-5G A137145-1000G
77272-316EA 10.3 USD
77272-316 77272-318 77272-320 77272-322 77747-971
Boc-Glu-OH 95%
Boc-Glu-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
MDL Number: MFCD00037297
CAS Number: 2419-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR