3,4,5-Trihydroxybenzaldehyde monohydrate 95%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trihydroxybenzaldehyde hydrate

A137088-1G A137088-5G
77272-312EA 57.68 USD
77272-312 77272-314
3,4,5-Trihydroxybenzaldehyde monohydrate 95%
3,4,5-Trihydroxybenzaldehyde monohydrate

Precaución: for Research Use Only

Formula: C₇H₆O₄·1H₂O
MW: 172.14 g/mol
MDL Number: MFCD00149124
CAS Number: 207742-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR