4,5-Dimethyl-1,3-thiazol-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dimethylthiazol-2-amine, 2-Amino-4,5-dimethyl-1,3-thiazole

A137005-1G A137005-250MG A137005-25G A137005-5G
77285-562EA 27.81 USD
77285-562 77285-564 77285-566 77285-568
4,5-Dimethyl-1,3-thiazol-2-amine 95%
4,5-Dimethyl-1,3-thiazol-2-amine

Precaución: for Research Use Only

Formula: C₅H₈N₂S
MW: 128.2 g/mol
MDL Number: MFCD00462425
CAS Number: 2289-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR