2-(1-Pyrazolyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1H-Pyrazol-1-yl)pyridine

A136729-1G A136729-250MG A136729-5G
77272-286EA 50.47 USD
77272-286 77272-288 77272-290
2-(1-Pyrazolyl)pyridine 98%
2-(1-Pyrazolyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MW: 145.17 g/mol
MDL Number: MFCD09033534
CAS Number: 25700-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR