N-Methyl-1-(4-nitrophenyl)methanamine 97%

Supplier: Ambeed
Warning

A136680-100G A136680-1G A136680-250MG A136680-25G A136680-5G A136680-10G
77258-686EA 1540.88 USD
77258-686 77258-688 77258-690 77258-692 77258-694 77747-969
N-Methyl-1-(4-nitrophenyl)methanamine 97%
N-Methyl-1-(4-nitrophenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.17 g/mol
MDL Number: MFCD00666471
CAS Number: 19499-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR