Isoquinoline-1-amine 97%

Supplier: Ambeed
Warning

Synonyms: Isoquinolin-1-amine, 1-Aminoisoquinoline

A136638-10G A136638-1G A136638-250MG A136638-25G A136638-5G
77258-676EA 93.73 USD
77258-676 77258-678 77258-680 77258-682 77258-684
Isoquinoline-1-amine 97%
Isoquinoline-1-amine

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
MDL Number: MFCD00024137
CAS Number: 1532-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR