3-Fluoro-4-nitrobenzotrifluoride 97%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-4-(trifluoromethyl)nitrobenzene, 2-Fluoro-1-nitro-4-(trifluoromethyl)benzene

A136557-100MG A136557-1G A136557-250MG A136557-5G
77272-274EA 7.21 USD
77272-274 77272-276 77272-278 77272-280
3-Fluoro-4-nitrobenzotrifluoride 97%
3-Fluoro-4-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MW: 209.098 g/mol
MDL Number: MFCD03840208
CAS Number: 402-12-0
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR