4-Amino-3-fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester

A136527-100MG A136527-10G A136527-1G A136527-250MG A136527-25G A136527-5G
77285-534EA 5.15 USD
77285-534 77285-536 77285-538 77285-540 77285-542 77285-544
4-Amino-3-fluorophenylboronic acid pinacol ester 97%
4-Amino-3-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
MDL Number: MFCD09033884
CAS Number: 819058-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR