(3aS,3a'S,8aR,8a'R)-2,2'-(Cyclopentane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A136513-100MG A136513-1G A136513-250MG A136513-5G
77258-668EA 57.68 USD
77258-668 77258-670 77258-672 77258-674
(3aS,3a'S,8aR,8a'R)-2,2'-(Cyclopentane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole) ≥97%, ee 99%
(3aS,3a'S,8aR,8a'R)-2,2'-(Cyclopentane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₂O₂
MW: 384.47 g/mol
MDL Number: MFCD31619111
CAS Number: 182122-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR