JNJ-31020028 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(4-(2-(Diethylamino)-2-oxo-1-phenylethyl)piperazin-1-yl)-3-fluorophenyl)-2-(pyridin-3-yl)benzamide

A136309-10MG A136309-1MG A136309-25MG A136309-50MG A136309-5MG
77234-732EA 40 USD
77234-732 77234-734 77234-736 77234-738 77234-740
JNJ-31020028 98%
JNJ-31020028

Precaución: for Research Use Only

Formula: C₃₄H₃₆FN₅O₂
MDL Number: MFCD18782744
CAS Number: 1094873-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR