4-Bromo-4'-(pentyloxy)-1,1'-biphenyl 97%

Supplier: Ambeed
Warning

A136223-10G A136223-1G A136223-250MG A136223-25G A136223-5G
77258-658EA 42 USD
77258-658 77258-660 77258-662 77258-664 77258-666
4-Bromo-4'-(pentyloxy)-1,1'-biphenyl 97%
4-Bromo-4'-(pentyloxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₇H₁₉BrO
MDL Number: MFCD00096777
CAS Number: 63619-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR