2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
A136207-10G
A136207-1G
A136207-25G
A136207-5G
A136207-250MG
77246-730EA
110.21
USD
77246-730
77246-732
77246-734
77246-736
77745-968
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 97%
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₉H₁₀N₂S MW: 178.26 g/mol |
MDL Number:
MFCD00128278 CAS Number: 4651-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...