FTI-277 hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: (S)-methyl 2-(5-(((R)-2-amino-3-mercaptopropyl)amino)-[1,1'-biphenyl]-2-ylcarboxamido)-4-(methylthio)butanoate hydrochloride

A136104-1MG A136104-2MG A136104-50MG A136104-5MG
77234-724EA 75.19 USD
77234-724 77234-726 77234-728 77234-730
FTI-277 hydrochloride 98%
FTI-277 hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₃₀ClN₃O₃S₂
MW: 484.0749 g/mol
MDL Number: MFCD28385886
CAS Number: 180977-34-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR