4-Nitropyrocatechol 95%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrocatechol, 4-Nitrobenzene-1,2-diol

A136048-10G A136048-1G A136048-5G A136048-25G
77285-514EA 33 USD
77285-514 77285-516 77285-518 77747-968
4-Nitropyrocatechol 95%
4-Nitropyrocatechol

Precaución: for Research Use Only

Formula: C₆H₅NO₄
MW: 155.11 g/mol
MDL Number: MFCD00007242
CAS Number: 3316-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR