4-Cyano-3-fluorophenyl-trans-4-(4-n-propylcyclohexyl)benzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-3-fluorophenyl 4-(trans-4-propylcyclohexyl)benzoate

A136030-10G A136030-1G A136030-250MG A136030-25G A136030-5G
77272-242EA 51.5 USD
77272-242 77272-244 77272-246 77272-248 77272-250
4-Cyano-3-fluorophenyl-trans-4-(4-n-propylcyclohexyl)benzoate 97%
4-Cyano-3-fluorophenyl-trans-4-(4-n-propylcyclohexyl)benzoate

Precaución: for Research Use Only

Formula: C₂₃H₂₄FNO₂
MW: 365.448 g/mol
MDL Number: MFCD11053365
CAS Number: 92118-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR