4-(Heptyloxy)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid

A136024-100G A136024-10G A136024-1G A136024-250MG A136024-25G A136024-5G
77285-502EA 207 USD
77285-502 77285-504 77285-506 77285-508 77285-510 77285-512
4-(Heptyloxy)phenylboronic acid 97%
4-(Heptyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁BO₃
MDL Number: MFCD04039169
CAS Number: 136370-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR