3,5-Dimethyl-1-phenyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dimethyl-1-phenylpyrazole, 3,5-Dimethyl-1-phenyl-1H-pyrazole
A135920-1G
A135920-25G
77272-238EA
19.57
USD
77272-238
77272-240
3,5-Dimethyl-1-phenyl-1H-pyrazole 97%
3,5-Dimethyl-1-phenyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂ MW: 172.23 g/mol |
MDL Number:
MFCD00020728 CAS Number: 1131-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...