5-Chloroquinoline 97%

Supplier: Ambeed
Warning

A135791-100MG A135791-10G
77258-638EA 5.15 USD
77258-638 77258-640
5-Chloroquinoline 97%
5-Chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₆ClN
MW: 163.61 g/mol
MDL Number: MFCD00239413
CAS Number: 635-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR