LMK-235 98%

Supplier: Ambeed
Warning

Synonyms: N-[6-(hydroxyamino)-6-oxohexoxy]-3,5-dimethylbenzamide, LMK-235, LMK235, N-((6-(Hydroxyamino)-6-oxohexyl)oxy)-3,5-dimethylbenzamide, LMK 235

A135784-10MG A135784-1MG A135784-5MG A135784-50MG
77234-706EA 66.95 USD
77234-706 77234-708 77234-710 77566-680
LMK-235 98%
LMK-235

Precaución: For research use only.

Formula: C₁₅H₂₂N₂O₄
MW: 294.35 g/mol
MDL Number: MFCD26522892
CAS Number: 1418033-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR