6-Chloro-2-fluoro-3-methylbenzaldehyde 99%

Supplier: Ambeed
Warning

A135727-1G A135727-250MG
77234-696EA 20 USD
77234-696 77234-698
6-Chloro-2-fluoro-3-methylbenzaldehyde 99%
6-Chloro-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MDL Number: MFCD01631370
CAS Number: 286474-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR