1-(2-Thenoyl)-3,3,3-trifluoroacetone 97%

Supplier: Ambeed
Warning

Synonyms: (TTA), Thenoyltrifluoroacetone, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione, TTFA, 2-Thenoyltrifluoroacetone, (2-Thenoyl)-ω,ω,ω-trifluoroacetone

A135642-100G A135642-10G A135642-1G A135642-5G A135642-25G
77272-226EA 35.02 USD
77272-226 77272-228 77272-230 77272-232 77298-802
1-(2-Thenoyl)-3,3,3-trifluoroacetone 97%
1-(2-Thenoyl)-3,3,3-trifluoroacetone

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
MDL Number: MFCD00005445
CAS Number: 326-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR